Anti-LOX Mouse Monoclonal Antibody [clone: OTI9G6]

Supplier: OriGene
ORIGTA500871EA 581 EUR
ORIGTA500871
Anti-LOX Mouse Monoclonal Antibody [clone: OTI9G6]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LOX (lysyl oxidase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR