Anti-PLEK Mouse Monoclonal Antibody [clone: OTI2F10]

Supplier: OriGene
ORIGTA500901EA 581 EUR
ORIGTA500901
Anti-PLEK Mouse Monoclonal Antibody [clone: OTI2F10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100

Type: Primary
Antigen: PLEK (pleckstrin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR