Anti-CISD1 Mouse Monoclonal Antibody [clone: OTI4G3]

Supplier: OriGene
ORIGTA500927EA 603 EUR
ORIGTA500927
Anti-CISD1 Mouse Monoclonal Antibody [clone: OTI4G3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: CISD1 (CDGSH iron sulfur domain 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR