Anti-FAHD2A Mouse Monoclonal Antibody [clone: OTI9D4]

Supplier: OriGene
ORIGTA500944EA 581 EUR
ORIGTA500944
Anti-FAHD2A Mouse Monoclonal Antibody [clone: OTI9D4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100

Type: Primary
Antigen: FAHD2A (fumarylacetoacetate hydrolase domain containing 2A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR