Anti-NIT2 Mouse Monoclonal Antibody [clone: OTI5A3]

Supplier: OriGene
ORIGTA501121EA 581 EUR
ORIGTA501121
Anti-NIT2 Mouse Monoclonal Antibody [clone: OTI5A3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: NIT2 (nitrilase family, member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR