Anti-PDE4A Mouse Monoclonal Antibody [clone: OTI1F5]

Supplier: OriGene
ORIGTA501153EA 603 EUR
ORIGTA501153
Anti-PDE4A Mouse Monoclonal Antibody [clone: OTI1F5]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PDE4A (phosphodiesterase 4A, cAMP-specific)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR