Anti-GPI Mouse Monoclonal Antibody [clone: OTI2D2]

Supplier: OriGene
ORIGTA501171EA 603 EUR
ORIGTA501171
Anti-GPI Mouse Monoclonal Antibody [clone: OTI2D2]
Antibodies
  • Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: GPI (glucose-6-phosphate isomerase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR