Anti-CYP1A2 Mouse Monoclonal Antibody [clone: OTI7D12]

Supplier: OriGene
TA501173
ORIGTA501173EA 616 EUR
ORIGTA501173
Anti-CYP1A2 Mouse Monoclonal Antibody [clone: OTI7D12]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CYP1A2 (cytochrome P450, family 1, subfamily A, polypeptide 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR