Anti-ACSBG1 Mouse Monoclonal Antibody [clone: OTI2B6]

Supplier: OriGene
ORIGTA501176EA 603 EUR
ORIGTA501176
Anti-ACSBG1 Mouse Monoclonal Antibody [clone: OTI2B6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: ACSBG1 (acyl-CoA synthetase bubblegum family member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR