Anti-ERCC1 Mouse Monoclonal Antibody [clone: OTI1H1]

Supplier: OriGene
ORIGTA501184EA 603 EUR
ORIGTA501184
Anti-ERCC1 Mouse Monoclonal Antibody [clone: OTI1H1]
Antibodies
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ERCC1 (excision repair cross-complementation group 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR