Anti-DDX39B Mouse Monoclonal Antibody [clone: OTI2C5]

Supplier: OriGene
ORIGTA501186EA 603 EUR
ORIGTA501186
Anti-DDX39B Mouse Monoclonal Antibody [clone: OTI2C5]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50

Type: Primary
Antigen: DDX39B (DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR