Anti-HSPB8 Mouse Monoclonal Antibody [clone: OTI1E3]

Supplier: OriGene
ORIGTA501210EA 608 EUR
ORIGTA501210
Anti-HSPB8 Mouse Monoclonal Antibody [clone: OTI1E3]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HSPB8 (heat shock 22kDa protein 8)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR