Anti-PTPRE Mouse Monoclonal Antibody [clone: OTI3H3]

Supplier: OriGene
ORIGTA501234EA 603 EUR
ORIGTA501234
Anti-PTPRE Mouse Monoclonal Antibody [clone: OTI3H3]
Antibodies
Recommended Dilutions: Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:50-100

Type: Primary
Antigen: PTPRE (protein tyrosine phosphatase, receptor type, E)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR