Anti-TACC3 Mouse Monoclonal Antibody [clone: OTI8B7]

Supplier: OriGene
ORIGTA501255EA 603 EUR
ORIGTA501255
Anti-TACC3 Mouse Monoclonal Antibody [clone: OTI8B7]
Antibodies
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TACC3 (transforming, acidic coiled-coil containing protein 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR