Anti-IGF2BP2 Mouse Monoclonal Antibody [clone: OTI4D10]

Supplier: OriGene
TA501275
ORIGTA501275EA 616 EUR
ORIGTA501275
Anti-IGF2BP2 Mouse Monoclonal Antibody [clone: OTI4D10]
Antibodies
  • Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: IGF2BP2 (insulin-like growth factor 2 mRNA binding protein 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR