Anti-USP5 Mouse Monoclonal Antibody [clone: OTI1A8]

Supplier: OriGene
ORIGTA501312EA 581 EUR
ORIGTA501312
Anti-USP5 Mouse Monoclonal Antibody [clone: OTI1A8]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: USP5 (ubiquitin specific peptidase 5 (isopeptidase T))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR