Anti-CD5 Mouse Monoclonal Antibody [clone: OTI6F7]

Supplier: OriGene
ORIGTA501334EA 603 EUR
ORIGTA501334
Anti-CD5 Mouse Monoclonal Antibody [clone: OTI6F7]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: CD5 (CD5 molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR