Anti-TBXAS1 Mouse Monoclonal Antibody [clone: OTI1A9]

Supplier: OriGene
ORIGTA501366EA 603 EUR
ORIGTA501366
Anti-TBXAS1 Mouse Monoclonal Antibody [clone: OTI1A9]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TBXAS1 (thromboxane A synthase 1 (platelet))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR