Anti-HID1 Mouse Monoclonal Antibody [clone: OTI2H6]

Supplier: OriGene
TA501370
ORIGTA501370EA 616 EUR
ORIGTA501370
Anti-HID1 Mouse Monoclonal Antibody [clone: OTI2H6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HID1 (HID1 domain containing)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR