Anti-IDO2 Mouse Monoclonal Antibody [clone: OTI1A4]

Supplier: OriGene
ORIGTA501378EA 586 EUR
ORIGTA501378
Anti-IDO2 Mouse Monoclonal Antibody [clone: OTI1A4]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: IDO2 (indoleamine 2,3-dioxygenase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR