Anti-DGKB Mouse Monoclonal Antibody [clone: OTI1F9]

Supplier: OriGene
ORIGTA501381EA 608 EUR
ORIGTA501381
Anti-DGKB Mouse Monoclonal Antibody [clone: OTI1F9]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: DGKB (diacylglycerol kinase, beta 90kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR