Anti-MOB1B Mouse Monoclonal Antibody [clone: OTI1H2]

Supplier: OriGene
ORIGTA501388EA 608 EUR
ORIGTA501388
Anti-MOB1B Mouse Monoclonal Antibody [clone: OTI1H2]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100; Immunochemistry: 1:150

Type: Primary
Antigen: MOB1B (MOB kinase activator 1B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR