Anti-COX6A1 Mouse Monoclonal Antibody [clone: OTI6H8]

Supplier: OriGene
TA501398
ORIGTA501398EA 616 EUR
ORIGTA501398
Anti-COX6A1 Mouse Monoclonal Antibody [clone: OTI6H8]
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: COX6A1 (cytochrome c oxidase subunit VIa polypeptide 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR