Anti-SHPRH Mouse Monoclonal Antibody [clone: OTI1D12]

Supplier: OriGene
ORIGTA501467EA 593 EUR
ORIGTA501467
Anti-SHPRH Mouse Monoclonal Antibody [clone: OTI1D12]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SHPRH (SNF2 histone linker PHD RING helicase, E3 ubiquitin protein ligase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR