Anti-PAICS Mouse Monoclonal Antibody [clone: OTI2C9]

Supplier: OriGene
ORIGTA501485EA 603 EUR
ORIGTA501485
Anti-PAICS Mouse Monoclonal Antibody [clone: OTI2C9]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PAICS (phosphoribosylaminoimidazole carboxylase, phosphoribosylaminoimidazole succinocarboxamide synthetase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR