Anti-SLC30A3 Mouse Monoclonal Antibody [clone: OTI2H6]

Supplier: OriGene
TA501498
ORIGTA501498EA 598 EUR
ORIGTA501498
Anti-SLC30A3 Mouse Monoclonal Antibody [clone: OTI2H6]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SLC30A3 (solute carrier family 30 (zinc transporter), member 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR