Anti-C16orf72 Mouse Monoclonal Antibody [clone: OTI2D1]

Supplier: OriGene
TA501514
ORIGTA501514EA 593 EUR
ORIGTA501514
Anti-C16orf72 Mouse Monoclonal Antibody [clone: OTI2D1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: C16orf72 (chromosome 16 open reading frame 72)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR