Anti-SH3GL1 Mouse Monoclonal Antibody [clone: OTI3F8]

Supplier: OriGene
ORIGTA501539EA 603 EUR
ORIGTA501539
Anti-SH3GL1 Mouse Monoclonal Antibody [clone: OTI3F8]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: SH3GL1 (SH3-domain GRB2-like 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR