Anti-TRIM2 Mouse Monoclonal Antibody [clone: OTI4E5]

Supplier: OriGene
ORIGTA501551EA 603 EUR
ORIGTA501551
Anti-TRIM2 Mouse Monoclonal Antibody [clone: OTI4E5]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: TRIM2 (tripartite motif containing 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR