Anti-NT5DC1 Mouse Monoclonal Antibody [clone: OTI1G10]

Supplier: OriGene
ORIGTA501593EA 603 EUR
ORIGTA501593
Anti-NT5DC1 Mouse Monoclonal Antibody [clone: OTI1G10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NT5DC1 (5'-nucleotidase domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR