Anti-VWA5A Mouse Monoclonal Antibody [clone: OTI3E10]

Supplier: OriGene
ORIGTA501636EA 603 EUR
ORIGTA501636
Anti-VWA5A Mouse Monoclonal Antibody [clone: OTI3E10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: VWA5A (von Willebrand factor A domain containing 5A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR