Anti-PRKD2 Mouse Monoclonal Antibody [clone: OTI3F7]

Supplier: OriGene
ORIGTA501647EA 603 EUR
ORIGTA501647
Anti-PRKD2 Mouse Monoclonal Antibody [clone: OTI3F7]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PRKD2 (protein kinase D2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR