Anti-RBBP9 Mouse Monoclonal Antibody [clone: OTI2G2]

Supplier: OriGene
TA501676
ORIGTA501676EA 616 EUR
ORIGTA501676
Anti-RBBP9 Mouse Monoclonal Antibody [clone: OTI2G2]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RBBP9 (retinoblastoma binding protein 9)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR