Anti-RIT2 Mouse Monoclonal Antibody [clone: OTI4B5]

Supplier: OriGene
ORIGTA501704EA 603 EUR
ORIGTA501704
Anti-RIT2 Mouse Monoclonal Antibody [clone: OTI4B5]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RIT2 (Ras-like without CAAX 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR