Anti-PTPN11 Mouse Monoclonal Antibody [clone: OTI3F2]

Supplier: OriGene
ORIGTA501758EA 608 EUR
ORIGTA501758
Anti-PTPN11 Mouse Monoclonal Antibody [clone: OTI3F2]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PTPN11 (protein tyrosine phosphatase, non-receptor type 11)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR