Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: OTI7B12]

Supplier: OriGene
TA501760
ORIGTA501760EA 593 EUR
ORIGTA501760
Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: OTI7B12]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PIK3AP1 (phosphoinositide-3-kinase adaptor protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR