Anti-HSPA6 Mouse Monoclonal Antibody [clone: OTI1E11]

Supplier: OriGene
ORIGTA501772EA 581 EUR
ORIGTA501772
Anti-HSPA6 Mouse Monoclonal Antibody [clone: OTI1E11]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HSPA6 (heat shock 70kDa protein 6 (HSP70B'))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR