Anti-PECR Mouse Monoclonal Antibody [clone: OTI1C9]

Supplier: OriGene
ORIGTA501809EA 603 EUR
ORIGTA501809
Anti-PECR Mouse Monoclonal Antibody [clone: OTI1C9]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PECR (peroxisomal trans-2-enoyl-CoA reductase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR