Anti-PUS7 Mouse Monoclonal Antibody [clone: OTI4H5]

Supplier: OriGene
TA501823
ORIGTA501823EA 616 EUR
ORIGTA501823
Anti-PUS7 Mouse Monoclonal Antibody [clone: OTI4H5]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PUS7 (pseudouridylate synthase 7 (putative))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR