Anti-ADI1 Mouse Monoclonal Antibody [clone: OTI5C2]

Supplier: OriGene
ORIGTA501827EA 608 EUR
ORIGTA501827
Anti-ADI1 Mouse Monoclonal Antibody [clone: OTI5C2]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: ADI1 (acireductone dioxygenase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR