Anti-AK3 Mouse Monoclonal Antibody [clone: OTI3G1]

Supplier: OriGene
ORIGTA501834EA 603 EUR
ORIGTA501834
Anti-AK3 Mouse Monoclonal Antibody [clone: OTI3G1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: AK3 (adenylate kinase 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR