Anti-PYCR2 Mouse Monoclonal Antibody [clone: OTI4A10]

Supplier: OriGene
ORIGTA501877EA 603 EUR
ORIGTA501877
Anti-PYCR2 Mouse Monoclonal Antibody [clone: OTI4A10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PYCR2 (pyrroline-5-carboxylate reductase family, member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR