Anti-GSTT2 Mouse Monoclonal Antibody [clone: OTI4F9]

Supplier: OriGene
TA501882
ORIGTA501882EA 598 EUR
ORIGTA501882
Anti-GSTT2 Mouse Monoclonal Antibody [clone: OTI4F9]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GSTT2 (glutathione S-transferase theta 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR