Anti-HSPA6 Mouse Monoclonal Antibody [clone: OTI1D12]

Supplier: OriGene
ORIGTA501950EA 593 EUR
ORIGTA501950
Anti-HSPA6 Mouse Monoclonal Antibody [clone: OTI1D12]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: HSPA6 (heat shock 70kDa protein 6 (HSP70B'))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR