Anti-SPR Mouse Monoclonal Antibody [clone: OTI2F7]

Supplier: OriGene
TA501982
ORIGTA501982EA 593 EUR
ORIGTA501982
Anti-SPR Mouse Monoclonal Antibody [clone: OTI2F7]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: SPR (sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR