Anti-SLFNL1 Mouse Monoclonal Antibody [clone: OTI5C3]

Supplier: OriGene
TA502017
ORIGTA502017EA 616 EUR
ORIGTA502017
Anti-SLFNL1 Mouse Monoclonal Antibody [clone: OTI5C3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SLFNL1 (schlafen-like 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR