Anti-TTL Mouse Monoclonal Antibody [clone: OTI3G5]

Supplier: OriGene
ORIGTA502027EA 586 EUR
ORIGTA502027
Anti-TTL Mouse Monoclonal Antibody [clone: OTI3G5]
Antibodies
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: TTL (tubulin tyrosine ligase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR