Anti-SNTA1 Mouse Monoclonal Antibody [clone: OTI2B9]

Supplier: OriGene
ORIGTA502043EA 603 EUR
ORIGTA502043
Anti-SNTA1 Mouse Monoclonal Antibody [clone: OTI2B9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SNTA1 (syntrophin, alpha 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR