Anti-RASD2 Mouse Monoclonal Antibody [clone: OTI1D11]

Supplier: OriGene
ORIGTA502073EA 603 EUR
ORIGTA502073
Anti-RASD2 Mouse Monoclonal Antibody [clone: OTI1D11]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: RASD2 (RASD family, member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR