Anti-CLPP Mouse Monoclonal Antibody [clone: OTI1D3]

Supplier: OriGene
ORIGTA502075EA 603 EUR
ORIGTA502075
Anti-CLPP Mouse Monoclonal Antibody [clone: OTI1D3]
Antibodies
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CLPP (caseinolytic mitochondrial matrix peptidase proteolytic subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR